Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
USP50
Entrez Gene ID
506271
Gene Name
ubiquitin specific peptidase 50
Gene Aliases
-
Chromosome Location
Chr.10: 59758049 - 59796155 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001080230.2 NP_001073699.2 3 - 4 590 143
GenBank mRNA BC102596.1 - 3 - 4 590 143

View Full Product Details