Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
COX8A
Entrez Gene ID
281091
Gene Name
cytochrome c oxidase subunit VIIIA (ubiquitous)
Gene Aliases
COX8, COX8L
Chromosome Location
Chr.29: 42308291 - 42310057 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_174024.2 NP_776449.2 1 - 2 177 75
GenBank mRNA BC108170.1 - 1 - 2 177 75
BC142020.1 - 1 - 2 144 75
J05201.1 - 1 - 2 119 75

View Full Product Details