Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
HSPA8
Entrez Gene ID
281831
Gene Name
heat shock protein family A (Hsp70) member 8
Gene Aliases
HSPA10, Hsc70
Chromosome Location
Chr.15: 33679011 - 33683436 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_174345.4 NP_776770.2 4 - 5 640 96
GenBank mRNA BC105182.1 - 4 - 5 641 96
BT030487.1 - 4 - 5 626 96
X53335.1 - - 565 96
X53827.1 - - 599 96

View Full Product Details