Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
HSPA8
Entrez Gene ID
281831
Gene Name
heat shock protein family A (Hsp70) member 8
Gene Aliases
HSPA10, Hsc70
Chromosome Location
Chr.15: 33679011 - 33683436 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_174345.4 NP_776770.2 5 - 6 1197 89
GenBank mRNA BC105182.1 - 5 - 6 1198 89
BC154389.1 - 1 - 2 315 89
BT030487.1 - 5 - 6 1183 89
X53335.1 - - 1122 89
X53827.1 - - 1156 89

View Full Product Details