Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
FAU
Entrez Gene ID
282847
Gene Name
FAU ubiquitin like and ribosomal protein S30 fusion
Gene Aliases
-
Chromosome Location
Chr.29: 43326668 - 43328299 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_174731.4 NP_777156.1 3 - 4 326 92
GenBank mRNA AF520959.1 - 3 - 4 249 92
BC102873.1 - 3 - 4 327 92

View Full Product Details