Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
EIF2S1
Entrez Gene ID
327694
Gene Name
eukaryotic translation initiation factor 2 subunit alpha
Gene Aliases
-
Chromosome Location
Chr.10: 79255235 - 79278260 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_175813.2 NP_787007.1 2 - 3 285 86
GenBank mRNA BC104590.1 - 2 - 3 285 86

View Full Product Details