The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_175813.2 | NP_787007.1 | 7 - 8 | 963 | 73 |
GenBank mRNA | BC104590.1 | - | 7 - 8 | 963 | 73 |
X53689.1 | - | - | 732 | 73 |