Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
DMAC2L
Entrez Gene ID
493709
Gene Name
distal membrane arm assembly complex 2 like
Gene Aliases
ATP5S, FB
Chromosome Location
Chr.10: 43219972 - 43232045 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001007812.3 NP_001007813.1 1 - 2 55 69
GenBank mRNA AY780292.1 - 1 - 2 44 69
BC109757.1 - 1 - 2 57 69

View Full Product Details