Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
HSF2BP
Entrez Gene ID
617142
Gene Name
heat shock transcription factor 2 binding protein
Gene Aliases
-
Chromosome Location
Chr.1: 143641658 - 143729079 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001035493.3 NP_001030570.1 6 - 7 893 72
GenBank mRNA BC110166.1 - 6 - 7 894 72
BT021887.1 - 6 - 7 906 72

View Full Product Details