Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
C3H1orf52
Entrez Gene ID
509947
Gene Name
chromosome 3 C1orf52 homolog
Gene Aliases
C1orf52
Chromosome Location
Chr.3: 58835524 - 58842785 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001037592.1 NP_001032681.1 1 - 2 728 74
GenBank mRNA BC109610.1 - 1 - 2 728 74

View Full Product Details