Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
NUTF2
Entrez Gene ID
614921
Gene Name
nuclear transport factor 2
Gene Aliases
-
Chromosome Location
Chr.18: 35354061 - 35370069 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001037624.1 NP_001032713.1 3 - 4 264 83
GenBank mRNA BC109983.1 - 3 - 4 264 83

View Full Product Details