Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
C15H11orf49
Entrez Gene ID
505437
Gene Name
chromosome 15 C11orf49 homolog
Gene Aliases
C11orf49
Chromosome Location
Chr.15: 76852567 - 77063233 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001038037.2 NP_001033126.1 7 - 8 814 59
GenBank mRNA BC109973.1 - 7 - 8 815 59

View Full Product Details