Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PPP1R32
Entrez Gene ID
506337
Gene Name
protein phosphatase 1 regulatory subunit 32
Gene Aliases
C11orf66, C29H11orf66
Chromosome Location
Chr.29: 39953298 - 39962271 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001038044.2 NP_001033133.1 6 - 7 922 110
GenBank mRNA BC111283.1 - 6 - 7 923 110

View Full Product Details