Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
KCTD13
Entrez Gene ID
507911
Gene Name
potassium channel tetramerization domain containing 13
Gene Aliases
-
Chromosome Location
Chr.25: 26343322 - 26356567 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001038057.2 NP_001033146.1 4 - 5 721 91
GenBank mRNA BC111242.1 - 4 - 5 722 91

View Full Product Details