Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TMCC3
Entrez Gene ID
509239
Gene Name
transmembrane and coiled-coil domain family 3
Gene Aliases
-
Chromosome Location
Chr.5: 24240322 - 24595453 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001038063.2 NP_001033152.1 2 - 3 1215 96
GenBank mRNA BC111266.1 - 2 - 3 1216 96

View Full Product Details