Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
BMI1
Entrez Gene ID
510666
Gene Name
BMI1 proto-oncogene, polycomb ring finger
Gene Aliases
PCGF4
Chromosome Location
Chr.13: 23430480 - 23441040 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001038072.1 NP_001033161.1 8 - 9 802 148
GenBank mRNA BC109871.1 - 8 - 9 802 148

View Full Product Details