Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
B4GALT4
Entrez Gene ID
511328
Gene Name
beta-1,4-galactosyltransferase 4
Gene Aliases
-
Chromosome Location
Chr.1: 64019129 - 64050092 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001038078.1 NP_001033167.1 6 - 7 1034 61
GenBank mRNA BC109604.1 - 6 - 7 1034 61

View Full Product Details