Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ZMYND12
Entrez Gene ID
512257
Gene Name
zinc finger MYND-type containing 12
Gene Aliases
-
Chromosome Location
Chr.3: 103720824 - 103755638 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001191293.2 NP_001178222.2 4 - 5 654 59
GenBank mRNA BC109818.1 - 4 - 5 616 59

View Full Product Details