Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
METTL26
Entrez Gene ID
514636
Gene Name
methyltransferase like 26
Gene Aliases
C25H16orf13
Chromosome Location
Chr.25: 544097 - 545876 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001038106.2 NP_001033195.1 3 - 4 595 141
GenBank mRNA BC109870.1 - 3 - 4 597 141

View Full Product Details