The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001038129.2 | NP_001033218.1 | 2 - 3 | 271 | 109 |
GenBank mRNA | BC111325.1 | - | 2 - 3 | 272 | 109 |