Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
EFCAB3
Entrez Gene ID
524320
Gene Name
EF-hand calcium binding domain 3
Gene Aliases
-
Chromosome Location
Chr.19: 46824857 - 46862874 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001038141.2 NP_001033230.1 4 - 5 523 106
GenBank mRNA BC111336.1 - 4 - 5 524 106

View Full Product Details