Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
UBL3
Entrez Gene ID
526950
Gene Name
ubiquitin like 3
Gene Aliases
-
Chromosome Location
Chr.12: 30806229 - 30854061 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001038144.1 NP_001033233.1 1 - 2 211 69
GenBank mRNA BC111119.1 - 1 - 2 211 69

View Full Product Details