Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TUBA3E
Entrez Gene ID
534900
Gene Name
tubulin, alpha 3e
Gene Aliases
TUBA3, TUBA3D
Chromosome Location
Chr.17: 72342835 - 72348558 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001038163.2 NP_001033252.1 1 - 2 78 50
GenBank mRNA BC110002.1 - 1 - 2 81 50

View Full Product Details