Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SHFL
Entrez Gene ID
539087
Gene Name
shiftless antiviral inhibitor of ribosomal frameshifting
Gene Aliases
C7H19orf66, RyDEN
Chromosome Location
Chr.7: 14650071 - 14654413 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001038181.1 NP_001033270.1 4 - 5 338 63
GenBank mRNA BC109821.1 - 4 - 5 338 63

View Full Product Details