Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ERI1
Entrez Gene ID
540728
Gene Name
exoribonuclease 1
Gene Aliases
THEX1
Chromosome Location
Chr.27: 25061506 - 25078779 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001038192.2 NP_001033281.1 2 - 3 442 105
GenBank mRNA BC109681.1 - 2 - 3 443 105
BT030668.1 - 2 - 3 412 105

View Full Product Details