Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ZMAT5
Entrez Gene ID
616095
Gene Name
zinc finger matrin-type 5
Gene Aliases
-
Chromosome Location
Chr.17: 68864264 - 68887043 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001038210.2 NP_001033299.1 4 - 5 411 101
GenBank mRNA BC111131.1 - 4 - 5 412 101

View Full Product Details