Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CCDC175
Entrez Gene ID
617691
Gene Name
coiled-coil domain containing 175
Gene Aliases
C10H14orf38
Chromosome Location
Chr.10: 71781846 - 71856290 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001038221.2 NP_001033310.1 6 - 7 889 122
GenBank mRNA BC111193.1 - 6 - 7 890 122

View Full Product Details