Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MND1
Entrez Gene ID
618239
Gene Name
meiotic nuclear divisions 1
Gene Aliases
-
Chromosome Location
Chr.17: 4218295 - 4297908 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001038223.2 NP_001033312.1 2 - 3 151 133
GenBank mRNA BC109806.1 - 2 - 3 152 133

View Full Product Details