Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
COIL
Entrez Gene ID
507580
Gene Name
coilin
Gene Aliases
-
Chromosome Location
Chr.19: 7765322 - 7786694 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001038512.2 NP_001033601.1 3 - 4 1461 86
GenBank mRNA BC109844.1 - 3 - 4 1462 86

View Full Product Details