Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CALR3
Entrez Gene ID
508555
Gene Name
calreticulin 3
Gene Aliases
-
Chromosome Location
Chr.7: 6520966 - 6546057 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001038514.2 NP_001033603.1 7 - 8 960 72
GenBank mRNA BC109750.1 - 7 - 8 961 72

View Full Product Details