Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SET
Entrez Gene ID
538444
Gene Name
SET nuclear proto-oncogene
Gene Aliases
-
Chromosome Location
Chr.11: 99200794 - 99206521 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001038564.1 NP_001033653.1 2 - 3 153 139
GenBank mRNA BC109772.1 - 2 - 3 153 139
BT030717.1 - 3 - 4 499 139

View Full Product Details