Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CCDC86
Entrez Gene ID
540913
Gene Name
coiled-coil domain containing 86
Gene Aliases
-
Chromosome Location
Chr.29: 37170153 - 37178885 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001038574.1 NP_001033663.1 3 - 4 967 55
GenBank mRNA BC109492.1 - 3 - 4 967 55

View Full Product Details