Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SKA2
Entrez Gene ID
615847
Gene Name
spindle and kinetochore associated complex subunit 2
Gene Aliases
FAM33A
Chromosome Location
Chr.19: 10093788 - 10124959 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001038579.2 NP_001033668.1 3 - 4 323 111
GenBank mRNA BC109479.1 - 3 - 4 324 111

View Full Product Details