Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
C29H11orf98
Entrez Gene ID
616423
Gene Name
chromosome 29 C11orf98 homolog
Gene Aliases
C29H11orf48
Chromosome Location
Chr.29: 40992849 - 40996236 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001038581.2 NP_001033670.1 2 - 3 201 91
GenBank mRNA BC109665.1 - 2 - 3 202 91

View Full Product Details