Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ELP4
Entrez Gene ID
614139
Gene Name
elongator acetyltransferase complex subunit 4
Gene Aliases
-
Chromosome Location
Chr.15: 62296501 - 62553953 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001038684.2 NP_001033773.1 8 - 9 1066 125
GenBank mRNA BC110206.1 - 8 - 9 1067 125

View Full Product Details