Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
MORN3
Entrez Gene ID
531482
Gene Name
MORN repeat containing 3
Gene Aliases
-
Chromosome Location
Chr.17: 53594179 - 53605592 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001040550.2 NP_001035640.1 4 - 5 838 54
GenBank mRNA BC110021.1 - 4 - 5 839 54

View Full Product Details