Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PDCL2
Entrez Gene ID
531753
Gene Name
phosducin like 2
Gene Aliases
-
Chromosome Location
Chr.6: 70988548 - 71024317 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001040551.2 NP_001035641.1 4 - 5 497 95
GenBank mRNA BC109499.1 - 4 - 5 498 95

View Full Product Details