Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
BMP4
Entrez Gene ID
407216
Gene Name
bone morphogenetic protein 4
Gene Aliases
-
Chromosome Location
Chr.10: 66518533 - 66532572 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001045877.1 NP_001039342.1 2 - 3 569 59
GenBank mRNA AY854957.1 - 1 - 2 311 59
BC105344.1 - 2 - 3 569 59
GU233961.1 - 1 - 2 369 59

View Full Product Details