Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SLBP
Entrez Gene ID
444868
Gene Name
stem-loop binding protein
Gene Aliases
-
Chromosome Location
Chr.6: 116841868 - 116857400 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001045880.2 NP_001039345.1 5 - 6 557 94
GenBank mRNA BC110146.1 - 5 - 6 558 94

View Full Product Details