Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
DIABLO
Entrez Gene ID
493999
Gene Name
diablo IAP-binding mitochondrial protein
Gene Aliases
SMAC
Chromosome Location
Chr.17: 53156230 - 53173972 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001045882.2 NP_001039347.1 2 - 3 215 79
GenBank mRNA BC111127.1 - 2 - 3 216 79

View Full Product Details