Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CPT2
Entrez Gene ID
504502
Gene Name
carnitine palmitoyltransferase 2
Gene Aliases
-
Chromosome Location
Chr.3: 93032986 - 93057158 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001045889.2 NP_001039354.1 2 - 3 347 137
GenBank mRNA BC105423.1 - 2 - 3 348 137

View Full Product Details