Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TAF1A
Entrez Gene ID
505051
Gene Name
TATA-box binding protein associated factor, RNA polymerase I subunit A
Gene Aliases
-
Chromosome Location
Chr.16: 26044163 - 26076233 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001045908.1 NP_001039373.1 8 - 9 1096 133
GenBank mRNA BC105540.1 - 8 - 9 1096 133

View Full Product Details