Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RBM4B
Entrez Gene ID
505756
Gene Name
RNA binding motif protein 4B
Gene Aliases
-
Chromosome Location
Chr.29: 44692147 - 44702027 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001045928.2 NP_001039393.1 3 - 4 1228 64
GenBank mRNA BC112497.1 - 3 - 4 1229 64

View Full Product Details