Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CD48
Entrez Gene ID
508386
Gene Name
CD48 molecule
Gene Aliases
-
Chromosome Location
Chr.3: 8902055 - 8932532 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001046002.1 NP_001039467.1 1 - 2 152 84
GenBank mRNA BC112825.1 - 1 - 2 152 84

View Full Product Details