Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PNPLA2
Entrez Gene ID
508493
Gene Name
patatin like phospholipase domain containing 2
Gene Aliases
ATGL
Chromosome Location
Chr.29: 50548339 - 50553859 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001046005.2 NP_001039470.1 4 - 5 833 65
GenBank mRNA BC112803.1 - 4 - 5 834 65
FJ798978.1 - 4 - 5 696 65
FJ897536.1 - 4 - 5 696 65

View Full Product Details