Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
DNAAF6
Entrez Gene ID
509340
Gene Name
dynein axonemal assembly factor 6
Gene Aliases
CXHXorf41, PIH1D3
Chromosome Location
Chr.X: 55825153 - 55873067 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001046029.2 NP_001039494.1 2 - 3 285 90
GenBank mRNA BC112467.1 - 2 - 3 286 90

View Full Product Details