Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SELENBP1
Entrez Gene ID
510154
Gene Name
selenium binding protein 1
Gene Aliases
MTO
Chromosome Location
Chr.3: 19442682 - 19451515 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001046048.1 NP_001039513.1 3 - 4 242 79
GenBank mRNA BC105545.1 - 3 - 4 242 79

View Full Product Details