Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
FAAP24
Entrez Gene ID
510793
Gene Name
FA core complex associated protein 24
Gene Aliases
C18H19orf40
Chromosome Location
Chr.18: 43398415 - 43401998 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001046062.2 NP_001039527.1 4 - 5 462 65
GenBank mRNA BC112837.1 - 4 - 5 463 65

View Full Product Details