Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
METTL1
Entrez Gene ID
511197
Gene Name
methyltransferase like 1
Gene Aliases
-
Chromosome Location
Chr.5: 55693253 - 55697229 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001046077.2 NP_001039542.1 4 - 5 587 71
GenBank mRNA BC110252.1 - 4 - 5 588 71
BT029912.1 - 4 - 5 602 71

View Full Product Details