Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TMEM248
Entrez Gene ID
511252
Gene Name
transmembrane protein 248
Gene Aliases
C25H7orf42, C7orf42
Chromosome Location
Chr.25: 28278394 - 28319481 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001046081.2 NP_001039546.1 1 - 2 200 138
GenBank mRNA BC110157.1 - 1 - 2 201 138

View Full Product Details